VWR® Kingston, Safety Goggles
Supplier: VWR Collection
VWRI111-1841EA 6.7 SGD
VWRI111-1841
VWR® Kingston, Safety Goggles
Goggles
Wrap-around modern style goggles, especially designed for small faces, provides excellent panoramic vision. The goggles consist of a PC lens, PVC frame and polyester/rubber headband.
  • High comfort with maximum protection
  • Fitted with indirect ventilation
  • Opalescent frame prevents light reflection
Certifications: Compliant with: EN 166:2001. Frame: EN166 3 4 BT CE. Lens: VWR® 1 BT.
Order Now
SPECIFICATIONS
Environmentally Preferable