VWR® Rome, Safety Spectacles
Supplier: VWR Collection
VWRI111-1842EA 9.2 SGD
VWRI111-1842
VWR® Rome, Safety Spectacles
Safety Glasses
Wrap-around protective eyewear offering a comfortable and custom fit all day long.
  • Adjustable temple length enables wearers to fit the eyewear to their facial profile
  • Lightweight, frameless design
  • Soft nose bridge for added comfort
  • Easy lens replacement
Certifications: Compliant with EN 166, EN 170; EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Order Now
SPECIFICATIONS
Environmentally Preferable