VWR® Amsterdam, Safety Spectacles
Supplier: VWR Collection
VWRI111-1843EA 7.6 SGD
VWRI111-1843
VWR® Amsterdam, Safety Spectacles
Safety Glasses
Wrap-around, single-lens safety glasses combining fashion, fit and function.
  • Extremely lightweight
  • Pantoscopic adjustment allows lens angle modification
  • Rubber tips for added comfort
Certifications: Compliant with EN 166, EN 170; EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Order Now
SPECIFICATIONS
Environmentally Preferable