VWR® Prague, Safety Spectacles
Supplier: VWR Collection
VWRI111-1846EA 17.8 SGD
VWRI111-1846
VWR® Prague, Safety Spectacles
Safety Glasses
These economical, compact cover glasses (OTG) fit over most prescription eyewear.
  • Pantoscopic adjustment allows lens angle modification
  • Adjustable temple length
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)