VWR® Budapest, Safety Spectacles
Supplier: VWR Collection
VWRI111-1847EA 10.5 SGD
VWRI111-1847
VWR® Budapest, Safety Spectacles
Safety Glasses
Wrap-around protective eyewear combining fashion, fit and function. One-piece PC lens and a lightweight, frameless design ensures a comfortable custom fit all day long.
  • Adjustable temple length enables wearers to fit the eyewear to their facial profile
  • Pantoscopic adjustment allows lens angle modification
  • Integrated side shield
  • Soft nose bridge for extra comfort
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Order Now
SPECIFICATIONS
Environmentally Preferable