VWR®, Examination Gloves, Latex

Supplier: VWR Collection
VWRI112-2959EA 23.7 SGD
VWRI112-2959 VWRI112-2960 VWRI112-2961 VWRI112-2962 VWRI112-2963
VWR®, Examination Gloves, Latex
Gloves
These ambidextrous examination gloves are suitable for use in non hazardous environments.

  • Non sterile, powder-free
  • Textured finish for good grip

Weight: 6 g

Length: 230 mm

Weight: 6 g

Colour: Natural
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR