VWR® Nitrile Extra Light, Nitrile Gloves

Supplier: VWR Collection
VWRI112-4193EA 0 SGD
VWRI112-4193 VWRI112-4197 VWRI112-4194 VWRI112-4195 VWRI112-4196
VWR® Nitrile Extra Light, Nitrile Gloves
Gloves
These thin, ambidextrous gloves are suitable for use in medical applications, food processing and for general lab requirements in non hazardous areas.

  • Non sterile, powder-free, textured fingertips
  • Very thin: Provide excellent elasticity for good wearer comfort
  • Free from phthalates/softeners and allergenic latex proteins; particularly skin friendly for users with latex allergy
  • Beaded cuffs for easier donning
  • Low thickness and textured fingertips for excellent grip

Certifications: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4; ASTM D6319, ASTM F1More Product Information
Order Now

SPECIFICATIONS

Model Nitrile Extra Light
Ambidextrous / hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex / latex-free Latex-free
Powdered / powder-free Powder-free
Cuff style Beaded
Cuff thickness 0,10 mm*
Finger thickness 0,14 mm*
Palm thickness 0,12 mm*
EN norm EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
Food contact approved +

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR