VWR®, Cut Resistant Gloves, PU Coating
Supplier: VWR Collection
VWRI112-5673EA 92.6 SGD
VWRI112-5673 VWRI112-5674 VWRI112-5675 VWRI112-5676 VWRI112-5677 VWRI112-5678
VWR®, Cut Resistant Gloves, PU Coating
Gloves
This VWR® glove with polyurethane coating provides general cut protection for the widest range of applications.
  • ANSI A2 Cut Protection (Level B)
  • UHMWPE/nylon/spandex composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a risk of cuts, including material handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A2 Cut Resistance. EN 388:2016 (4X42B)
Certified by OKEO-TEX meeting Standard 100 certifying that no harmfulMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Latex / latex-free Latex-free
Finish PU palm coating
Gauge 13
Cuff style Knit wrist
EN norm EN 388:2016 (4X42B)