VWR® Maximum, Cut Resistant Gloves
Supplier: VWR Collection
VWRI112-5691EA 153 SGD
VWRI112-5691 VWRI112-5692 VWRI112-5693 VWRI112-5694 VWRI112-5695 VWRI112-5696
VWR® Maximum, Cut Resistant Gloves
Gloves
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used inMore Product Information