VWR®, Precision Grip Gloves, PU Coating
Supplier: VWR Collection
VWRI112-5697EA 29.1 SGD
VWRI112-5697 VWRI112-5698 VWRI112-5699 VWRI112-5726 VWRI112-5727 VWRI112-5728
VWR®, Precision Grip Gloves, PU Coating
Gloves
VWR® precision grip gloves with polyurethane coating provide basic cut protection and protection against mechanical risks.
  • ANSI A1 cut protection (Level A)
  • Nylon composition
  • Polyurethane (PU) coating on palm and fingertips
  • Black knitted glove with black coating
Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment. Also great for package handling, picking and packing.
Certifications: Meets requirements for ANSI A1 Cut Resistance. EN 388:2016 (4131X)
Certified by OKEO-TEX meeting Standard 100More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Latex / latex-free Latex-free
Finish PU palm coating
Gauge 15
Cuff style Knit wrist
EN norm EN 388:2016 (4131X)