VWR®, Precision Grip Gloves with Dots
Supplier: VWR Collection
VWRI112-5730EA 92.6 SGD
VWRI112-5730 VWRI112-5731 VWRI112-5732 VWRI112-5733 VWRI112-5734 VWRI112-5729
VWR®, Precision Grip Gloves with Dots
Gloves
This VWR® precision grip glove with grip dots and nitrile coating provides basic cut protection and enhanced grip.
  • ANSI A1 Cut Protection (Level A)
  • Grip dots on the palm for enhanced grip
  • Nylon composition
  • Nitrile (NBR) coating on palm and fingertips
  • Grey knitted glove with black coating
Ideal for handling, picking, packaging, automotive, or situations that require dexterity in a dry and/or dirty environment.
Certifications: Meets requirements for ANSI A1 Cut Resistance. EN 388:2016 (4131X)
Certified by OKEO-TEX meeting Standard 100 certifying thatMore Product Information