VWR® Cleanroom Gloves, Nitrile, Solid White, Non-Tacky, Class 10 / ISO 4
Supplier: VWR Collection
VWRI112-6199EA 389 SGD
VWRI112-6199 VWRI112-6200 VWRI112-6201 VWRI112-6202 VWRI112-6203 VWRI112-6204
VWR® Cleanroom Gloves, Nitrile, Solid White, Non-Tacky, Class 10 / ISO 4
Gloves
VWR® cleanroom nitrile gloves are manufactured from a synthetic copolymer of acrylonitrile, butadiene and methylacrylic acid, that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Beaded cuff
  • Nitrile
  • Silicone-free
  • Low particle count
Certifications: N ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Packaging: 100 gloves per double bag, 10 bags per case.