VWR® FFP2, Disposable Particle Filtering Respirators
Supplier: VWR Collection
VWRI113-0645EA 34.5 SGD
VWRI113-0645 VWRI113-0646
VWR® FFP2, Disposable Particle Filtering Respirators
Respirators Disposable Respirators Particulate Respirators
These masks provide protection against particulates, dust and aerosols. For use against non toxic to low toxicity solid and liquid aerosols.
  • Exhalation valve fitted with the exclusive `butterfly´ opening-closing system
  • Extra light
  • Latex- and PVC-free
EN 149:2001; EG 89/686 (11.B)
Certifications: EN 149:2001+A1:2009
Caution: *One size fits all