VWR® Maximum, Cleanroom Face Masks
Supplier: VWR Collection
VWRI113-6220EA 106 SGD
VWRI113-6220 VWRI113-6221 VWRI113-6222 VWRI113-9930 VWRI113-9931
VWR® Maximum, Cleanroom Face Masks
Masks Cleanroom Masks
Maximum cleanroom masks feature three ply construction for superior particle and bacterial filtration efficiency. These masks are latex-free.
  • Three ply masks provide high bacterial and particle filtration efficiency
  • Highly breathable and fluid resistant
  • A full length plastic arch holds the fabric away from the wearer’s mouth to allow cooler, more comfortable breathing
  • Malleable noseband ensures a custom, secure seal
  • Available with spandex ear loops or PP ties, ultrasonically welded to maintain softness and guard against particulates
  • Moisture resistant,
More Product Information