VWR®, Sleeve Protectors, PP/PE
Supplier: VWR Collection
VWRI113-8255EA 22.4 SGD
VWRI113-8255
VWR®, Sleeve Protectors, PP/PE
Disposable Garments
Sleeve protector made from non woven PP coated with PE.
  • Waterproof
  • Welded elasticated cuffs
Ø×L: 200×400 mm
Density: 48 g/m²