VWR® Basic ESD Bouffant Caps
Supplier: VWR Collection
VWRI113-8900EA 92.5 SGD
VWRI113-8900
VWR® Basic ESD Bouffant Caps
Disposable Garments
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Anti-static properties
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.