VWR® Basic ESD Visitor Coats, SPP
Supplier: VWR Collection
VWRI113-8954EA 130 SGD
VWRI113-8954
VWR® Basic ESD Visitor Coats, SPP
Disposable Garments
Disposable coats, with collar, made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Press stud closure
  • Elastic wrists
  • Free from natural rubber latex
  • Anti-static properties
Weight: 30 g/m²
Packaging: 1 piece per bag, 50 bags per case.