VWR® Basic Visitor Coats, PE
Supplier: VWR Collection
VWRI113-9092EA 519 SGD
VWRI113-9092
VWR® Basic Visitor Coats, PE
Disposable Garments
Disposable visitor coat made of PE film.
  • Hood and press stud closure
  • Free from natural rubber latex
Thickness: 15 µm
One size fits all
Packaging: 50 pieces per bag, 10 bags per case.