VWR®, Cleanroom Face Masks, Class 100
Supplier: VWR Collection
VWRI113-9756EA 41 SGD
VWRI113-9756 VWRI114-9345
VWR®, Cleanroom Face Masks, Class 100
Masks Cleanroom Masks
Cleanroom face masks are manufactured with non woven polypropylene/polyethylene (ES) / melt-blown polypropylene (MBPP) materials.
  • Low particle generation - suitable for critical environments
  • High bacterial filtration efficiency and low breathable resistance
  • Packed in a cleanroom environment
  • Solvent-safe double-bag cleanroom packaging
  • 30% nylon/70% polyester ear loops
  • Plastic wire nose clip
  • For industrial and cleanroom use only; do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of
More Product Information