VWR® Spec-Wipe® 4, Cleanroom Wipes, Sterile
Supplier: VWR Collection
VWRI115-1724EA 438 SGD
VWRI115-1724
VWR® Spec-Wipe® 4, Cleanroom Wipes, Sterile
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
These wipes are made from double-woven polyester fabric, with close-cut edges, and are washed in an ISO Class 4 cleanroom to minimise contamination by particles and extractables.
  • For critical process cleaning, and environments where sterility, low particle and fibre generation are critical
  • Gamma sterilised to SAL 10⁻⁶ (Sterility Assurance Level)
  • Resealable pouches to protect wipes from contamination
Ideal for cleaning abrasive surfaces, yet still soft enough for use on sensitive surfaces.
Suitable for use in Grade A, ISO 4 cleanrooms.