VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
VWRI116-0282EA 36.6 SGD
VWRI116-0282 VWRI116-0283 VWRI116-0285 VWRI116-0286 VWRI116-0281 VWRI116-0280 VWRI116-0279 VWRI116-0287 VWRI116-0284
VWR® Labels, Hazardous Substance, GHS Labels
Labels
These labels, made of PE film and with the GHS symbols, meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures. These GHS symbols replaced traditional orange symbols (HSID) in 2015, when the pictogram on an orange square and R and S phrases disappeared. They are based on the physical, health and environmental risks posed by the substance.
  • Diamond shape
  • Black print on white background in red frame
  • Roll contains 500 labels in a dispensing box
Order Now
SPECIFICATIONS
Environmentally Preferable