VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Supplier: VWR Collection
VWRI121-0137EA 353 SGD
VWRI121-0137 VWRI121-0143
VWR®, Absorbents - Mats, Rolls and Socks for Oil Spills
Spill Control Materials
VWR® absorbents and spill-containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
  • Will only absorb oil and petroleum-based liquids
  • Will not absorb water-based liquids
  • Can be used outdoors as they will not absorb water
Colour: White