VWR®, Absorbents - Mats, Rolls and Socks, for Oil, Water and Chemical Based Spills

Supplier: VWR Collection
VWRI121-0138EA 373 SGD
VWRI121-0138 VWRI121-0141 VWRI121-0144
VWR®, Absorbents - Mats, Rolls and Socks, for Oil, Water and Chemical Based Spills
Spill Control Materials
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.

  • Absorbs a variety of industrial liquids including oil, water, solvents and coolants
  • Cannot be left outdoors as they will absorb water
  • Maintenance and universal sorbents

For use with chemicals, oil and water.

Colour: Grey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR