VWR®, Absorbents - Mats, Rolls and Socks, for Chemical Based Spills
Supplier: VWR Collection
VWRI121-0142EA 175 SGD
VWRI121-0142 VWRI121-0145 VWRI121-0139
VWR®, Absorbents - Mats, Rolls and Socks, for Chemical Based Spills
Spill Control Materials
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
  • Absorbs a wide range of chemicals, including acids and caustics
  • Chemically inert: Product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid
  • Green colour alerts that product is being used for hazardous liquids
  • Green colour allows user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Cannot be left outdoors as they will absorb water
More Product Information