VWR®, Non Hazardous Waste Bags
Supplier: VWR Collection
VWRI129-0022EA 668 SGD
VWRI129-0022
VWR®, Non Hazardous Waste Bags
Bags Waste Disposal Bags
Sturdy bags, ideal for disposing of non hazardous waste that must first be autoclaved. In PP, 50 µm thick, transparent, with labelling area.
  • Autoclavable up to 121 °C
  • Highly resistant to tears and impact
  • Specially welded seams reduce risk of leakage
The bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709-98.
Certifications: Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709-98.