VWR®, Biohazard Bags
Supplier: VWR Collection
14220-084 14220-082
VWRI129-0030EA 412 SGD
VWRI129-0030 VWRI129-0032 VWRU14220-084 VWRU14220-082
VWR®, Biohazard Bags
Bags Waste Disposal Bags
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 75 µm thick, orange, with 'Biohazard' label.
  • Autoclavable up to 135 °C
  • The useful temperature indicator text is white before autoclaving, then black
  • 'Contents Have Been Autoclaved' indicates successful autoclaving
  • Luminous red for easy recognition and highly visible 'Biohazard' label with the corresponding warning sign
These bags are DOT approved.
Order Now
SPECIFICATIONS
Environmentally Preferable