VWR®, Biohazard Bags
Supplier: VWR Collection
VWRI129-0039EA 161 SGD
VWRI129-0039 VWRI129-0040
VWR®, Biohazard Bags
Bags Waste Disposal Bags
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Supplied complete with the necessary sealing wires. HDPE, 100 µm thick, orange, with 'Biohazard' label.
  • Autoclavable up to 121 °C
  • Highly resistant to tears and impact
  • Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign
Order Now
SPECIFICATIONS
Environmentally Preferable