VWR®, Zip-Seal Bags
Supplier: VWR Collection
VWRI129-0320EA 27.6 SGD
VWRI129-0320 VWRI129-0321 VWRI129-0322 VWRI129-0323 VWRI129-0324 VWRI129-0325 VWRI129-0293 VWRI129-0294 VWRI129-0295 VWRI129-0296 VWRI129-0297 VWRI129-0298 VWRI129-0299 VWRI129-0306 VWRI129-0307 VWRI129-0308 VWRI129-0309 VWRI129-0310 VWRI129-0311 VWRI129-0312 VWRI129-0313 VWRI129-0314 VWRI129-0315 VWRI129-0316 VWRI129-0317 VWRI129-0318 VWRI129-0319
VWR®, Zip-Seal Bags
Bags Sample Bags
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
  • Quick sealing with guide tracks on the pressure seal
  • Transparent PE for easy identification of the contents
  • Can be resealed repeatedly, flexible and tear-proof
Packaging: 100/bag, 10 bags/case.
Order Now
SPECIFICATIONS
Environmentally Preferable