VWR® Disposal Bags
Supplier: VWR Collection
VWRI129-0582EA 293 SGD
VWRI129-0582 VWRI129-0583 VWRI129-0578 VWRI129-0584 VWRI129-0585 VWRI129-0586 VWRI129-0587 VWRI129-0588 VWRI129-0589 VWRI129-0579 VWRI129-0580 VWRI129-0581
VWR® Disposal Bags
Bags Waste Disposal Bags
For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Biohazard' printing.
  • Heat resistant up to 145 °C
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable