VWR® Blender Bags
Supplier: VWR Collection
VWRI129-0728EA 78.3 SGD
VWRI129-0728 VWRI129-0729 VWRI129-0730 VWRI129-0731 VWRI129-0732 VWRI129-0733 VWRI129-0734
VWR® Blender Bags
Bags Blender Bags
Wide choice of PE sterile bags from 80 to 3500 ml capacity. Fit all conventional laboratory blenders, adapted to all samples.
  • Bags with filters are mainly dedicated to fibrous samples
  • Bags with full filter for pasty samples
  • Plain bags for standard uses
  • No risk of cross-contamination
Absence of heavy material : raw material is compliant with the EC/94/62 directive.
Food contact: raw material is compliant with regulation EC N°1935/2004
Based on ISO 11137: Sterilisation of health care products SAL10-3
Certifications: Absence of heavy material: RawMore Product Information