VWR®, High Performance Blender Bags
Supplier: VWR Collection
VWRI129-1102EA 121 SGD
VWRI129-1102 VWRI129-1103 VWRI129-1104 VWRI129-1105 VWRI129-1106
VWR®, High Performance Blender Bags
Bags Blender Bags
Complete range of bags, sterile and alimentary certified, for safe and quick preparation of solid samples before microbiological and chemical analysis. Bags are resistant for three minutes blending (frequency 4 t/second). They fit all conventional 400 ml laboratory blenders, and are adapted to all samples.
  • Can resist to high temperature from –40 to +80 °C
  • Can be frozen
  • Gamma irradiated: No risk of cross-contamination (the red sticker on every box serves as a qualitative indicator of gamma ray processing)
  • Colour-change label when sterile: Red colour, for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable