VWR®, Autoclavable Biohazard Bags
Supplier: VWR Collection
VWRI129-1148EA 315 SGD
VWRI129-1148 VWRI129-1151 VWRI129-1152
VWR®, Autoclavable Biohazard Bags
Bags Waste Disposal Bags
Ideal for processing biohazardous materials that require autoclaving prior to disposal. Available with or without temperature indicator. High integrity polypropylene bags withstand 135 °C (275 °F) maximum autoclave temperature. PP, 50 µm thick, yellow.
  • Twist-tie closure allows bag to expand and vent steam during cooling
  • Bags resist punctures, tears and leaks, and they also meet the 165 g dart-drop standard as required by ASTM D1709-98
  • Each bag is imprinted with the universal biohazard symbol
All biohazard bags have a temperature indicator patch withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable