VWR®, Biodegradable Bags, Twist-Seal
Supplier: VWR Collection
VWRI129-1488EA 856 SGD
VWRI129-1488 VWRI129-1516 VWRI129-1538 VWRI129-1541 VWRI129-1543 VWRI129-1546
VWR®, Biodegradable Bags, Twist-Seal
Bags Sample Bags
LDPE bags with safety tabs, manufactured and packed in sterile conditions.
  • Provides a secure, contaminant-free, flexible container that ensures dependandle analytical results
  • RNase-, DNase-, pyrogen- and BPA-free
  • Offers an economical and efficient way to collect,contain and carry samples
  • Wire tab keeps bag open for filling
  • Unique double fold over system provides a temporary leakproof seal
Made of highly resistant virgin LDPE containing additives that promote in landfill conditions. The biodegradation does not start before it is put in landfillMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable