VWR®, Incineration Bags
Supplier: VWR Collection
14220-112
VWRU14220-112EA 666 SGD
VWRU14220-112 VWRI129-1569
VWR®, Incineration Bags
Bags Waste Disposal Bags
Constructed of durable PP with feature the universal biohazard symbol in black.
Certifications: Meets the 165 g dart drop standard performed per ASTM D1709-98 and the 480 g tear standard performed per ASTM D1299-94A.
Caution: Autoclaving not recommended. Bags will melt at 98,9 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable