BinLiners™ and BinLiner™ Stands
Supplier: MICRONOVA
BL2-40TRIR BL2-40TRIRB BL4-40TRIRB BL4-40TRIRBB BL4-40TRIR BL1-40TRIRBB
MICRBL2-40TRIREA 0 SGD
MICRBL2-40TRIR MICRBL2-40TRIRB MICRBL4-40TRIRB MICRBL4-40TRIRBB MICRBL4-40TRIR MICRBL1-40TRIRBB VWRI129-3991
BinLiners™ and BinLiner™ Stands
Bags Waste Disposal Bags
LDPE Micronova BinLiners™ are ideal for the collection and disposal of waste, collection and transportation of parts or raw materials, and storage in cleanrooms and controlled environments. Thickness of 1 to 6 mm available for a variety of applications; including covering equipment, holding production waste, document storage, lining large production totes and so on.
  • Made with FDA compliant material
  • Single-seam construction
  • Gusseted base seals for a better fit in rectangular containers
  • Plastic core roll pack and flat pack options
  • Available in multiple sizes
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable