VWR® Autoclave Bags, Polypropylene

Supplier: VWR Collection
VWRI129-4026EA 225 SGD
VWRI129-4026 VWRI129-4027 VWRI129-4030 VWRI129-4031 VWRI129-4032 VWRI129-4033 VWRI129-4034 VWRI129-4035 VWRI129-4036 VWRI129-4038 VWRI129-4039 VWRI129-4040 VWRI129-4042 VWRI129-4043
VWR® Autoclave Bags, Polypropylene
Bags Waste Disposal Bags

Sturdy, temperature resistant bags, ideal for disposing of non-hazardous waste or hazardous waste that must first be autoclaved. Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.



  • Autoclavable up to 135 °C maximum

  • Include 200 mm twist ties which allow bag to expand and vent steam during cooling

  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Available in different thickness: 37,5, 50  or 76 µm thick
  • Available
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR