VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
VWRI129-4027EA 182 SGD
VWRI129-4027 VWRI129-4030 VWRI129-4032 VWRI129-4033 VWRI129-4038 VWRI129-4039 VWRI129-4040 VWRI129-4043
VWR® Autoclavable Bags, Polypropylene, Clear
Bags Waste Disposal Bags
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste that must first be autoclaved.
  • Autoclavable up to 135 °C (275 °F) maximum
  • 20 cm twist ties included allows bag to expand and vent steam during cooling
  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Available with differnent thickness 37,5 µm, 50 µm thick
  • Available with plain or printed bag
  • Highly resistant to tears and punctures
  • Without autoclave Indicator
Can also be used for autoclaving other products. Supplied complete
More Product Information