VWR®, Sterile Cleanroom Double-Pouch Twist-Seal Bags
Supplier: VWR Collection
VWRI129-9889EA 1280 SGD
VWRI129-9889 VWRI129-9890 VWRI129-0665 VWRI129-9000 VWRI129-9003 VWRI129-9001 VWRI129-9002 VWRI129-9004 VWRI129-9005 VWRI129-9006 VWRI129-9007 VWRI129-9008 VWRI129-9009 VWRI129-9010 VWRI129-9011 VWRI129-9012 VWRI129-9013 VWRI129-9014 VWRI129-9015 VWRI129-9016 VWRI129-9017 VWRI129-9018
VWR®, Sterile Cleanroom Double-Pouch Twist-Seal Bags
Bags Sample Bags
PE bags, transparent. These sterile sampling bags provide a secure, contaminant-free pliant container that ensures dependable analysis results. Economical and efficient way to collect, contain and carry samples. With these bags, the user can bring sterile sampling bags into a cleanroom without any worries about contamination. The bags are double-pouched so the user can remove the outer bag before entering the cleanroom, which minimises the number of bags exposed.
  • The bags are sterilised inside and out by gamma irradiation
  • Sterility certificate in every box
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable