VWR®, Twist-Seal Bags
Supplier: VWR Collection
VWRI129-9854EA 239 SGD
VWRI129-9854 VWRI129-0012 VWRI129-0013 VWRI129-0010 VWRI129-0011 VWRI129-9870 VWRI129-9884 VWRI129-9885 VWRI129-9886 VWRI129-9887 VWRI129-0417 VWRI129-0418 VWRI300-0107 VWRI129-9019 VWRI129-9020 VWRI129-9021 VWRI129-9022 VWRI129-9023 VWRI129-9024 VWRI129-9025 VWRI129-9026 VWRI129-9027 VWRI129-9028 VWRI129-1161 VWRI129-1162 VWRI129-9800 VWRI129-9801 VWRI129-9802 VWRI129-9803 VWRI129-9804 VWRI129-9805 VWRI129-9806 VWRI129-9808 VWRI129-9807 VWRI129-9809 VWRI129-9810 VWRI129-9813 VWRI129-9814 VWRI129-9815 VWRI129-9816 VWRI129-9817 VWRI129-9029
VWR®, Twist-Seal Bags
Bags Sample Bags
LDPE bags with safety tabs, manufactured and packed in sterile conditions.
  • Provides a secure, contaminant-free, flexible container that ensures dependable analytical results
  • RNase-, DNase-, pyrogen- and BPA-free
  • Offers an economical and efficient way to collect, contain and carry samples
  • Wire tab keeps bag open for filling
  • Unique double fold-over system provides a temporary leakproof seal
Universal use, for samples such as food, liquid, grain, petroleum products, animal material, waste and soil. Suitable for environmental sampling (surface samples),More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable