VWR®, Bags, Blue Twist-Seal
Supplier: VWR Collection
VWRI129-9608EA 822 SGD
VWRI129-9608 VWRI129-9609 VWRI129-9614
VWR®, Bags, Blue Twist-Seal
Bags Sample Bags
This line of blue twist-seal printed bags, with safety tab and two round wires, has been specifically designed for food industry. The bags accommodate solid, semi-solid and liquid compounds.
  • Blue tear-off for increased visibility
  • Distinct colour avoids possible foreign matter contamination
  • Visual recognition of the product, but can also be used to segregate within a factory environment
  • Made from virgin PE
  • Optimised opening area to facilitate insertion of sample
  • Lot number traceability
  • Bags accommodate solid, semi-solid and liquid compounds
  • All sterile
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable