VWR® Safety Shower Testing Unit
Supplier: VWR Collection
VWRI139-0806EA 4320 SGD
VWRI139-0806
VWR® Safety Shower Testing Unit
First Aid Supplies Showers
Laboratory safety showers should be tested once a month to check the functionality and the required water flow rate of 60 L/min. for emergency instances. This mobile test unit provides a very convenient and easy solution to test both body safety showers and handheld eye showers.
  • Transparent PVC water reservoir on castors
  • 110 litre capacity
  • Built-in suction pump (230 V) and IP 45 protected power switch
  • Drain hose (2 m) to empty the water reservoir
  • Separate mounts for drain hose and power cord
Placed centrally under the body safety shower, the largeMore Product Information