VWR® Bucket Liners, Sterile
Supplier: VWR Collection
VWRI141-1380EA 564 SGD
VWRI141-1380 VWRI141-1443
VWR® Bucket Liners, Sterile
Bags Waste Disposal Bags
Bucket liners create an impermeable barrier between the disinfectant and receptacle.
  • LDPE/nylon blend
  • Irradiated
  • Fits B-7 series buckets
Helps ensure the integrity of a disinfectant blend.
Packaging: Individually bagged, double-bagged in groups of 6. 4 packs of 6 per case.
Order Now
SPECIFICATIONS
Environmentally Preferable