VWR® LED and LCD Ultrasonic Cleaning Baths
Supplier: VWR Collection
VWRI142-0168EA 727 SGD
VWRI142-0168 VWRI142-0169 VWRI142-0170 VWRI142-0171 VWRI142-0172 VWRI142-0173 VWRI142-0174 VWRI142-0175 VWRI142-0176 VWRI142-0177 VWRI142-0178 VWRI142-0179 VWRI142-0180 VWRI142-0181 VWRI142-0182 VWRI142-0183 VWRI142-0184 VWRI142-0185 VWRI142-0186 VWRI142-0187 VWRI142-0188 VWRI142-0189 VWRI142-0190 VWRI142-0191 VWRI142-0192 VWRI142-0193
VWR® LED and LCD Ultrasonic Cleaning Baths
Ultrasonic Baths
The housing and tank are constructed from high-quality SUS 304 stainless steel, providing durability and longevity. Equipped with user-friendly interface display which offers intuitive function selection and real-time parameter setting. Units also feature intelligent detection on startup, promptly alerting users to any potential issues via buzzer and alarm. Additionally, they continuously monitor the power supply voltage in real-time to ensure optimal operating conditions. The automatic heating stirring function ensures uniform liquid temperature throughout the tank. The anti-leakage trayMore Product Information