VWR® Critical Swabs
Supplier: VWR Collection
89031-276 89031-286
VWRI149-0334EA 565 SGD
VWRI149-0334 VWRI149-0339 VWRU89031-276 VWRU89031-286
VWR® Critical Swabs
Swabs Swabs for Sampling
Swabs feature 100 ppi open or closed cell polyurethane foam, with 100% virgin polypropylene handle. Some have a blue, glass fibre, reinforced polypropylene shaft for extra rigidity, especially for use in critical environments.
  • Non abrasive to protect delicate components
  • Heads are thermally-bonded to the handle without adhesive
  • Withstand most widely used solvents in controlled environments
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904