VWR®, Ultrapure Water System, U Series
Supplier: VWR Collection
VWRI171-1097EA 10570 SGD
VWRI171-1097 VWRI171-1246 VWRI171-1098 VWRI171-1099 VWRI171-1244 VWRI171-1245 VWRI171-1467 VWRI171-1468 VWRI171-1469 VWRI171-1470 VWRI171-1471 VWRI171-1472
VWR®, Ultrapure Water System, U Series
Water Purification Systems Pure and Ultra Pure Water Systems
This 'two in one' lab water system produces ultrapure (Type I) and RO (Type III) water from tap water. Quality of ultrapure water meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user with full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
  • RFID-tagged consumables
More Product Information