VWR®, Ultrapure Water System, P Series
Supplier: VWR Collection
VWRI171-1248EA 14590 SGD
VWRI171-1248 VWRI171-1247 VWRI171-1462 VWRI171-1463
VWR®, Ultrapure Water System, P Series
Water Purification Systems Pure and Ultra Pure Water Systems
This powerful and intuitive laboratory water purification system produces Type I ultrapure water from RO, distilled or deionised water. The product water quality meets or exceeds CLSI and CAP Type I water standards.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
  • RFID-tagged
More Product Information