VWR® VCP 80/VCP 130 Vacuum/Pressure Pumps

Supplier: VWR Collection
VWRI181-0308EA 1400 SGD
VWRI181-0308 VWRI181-0309
VWR® VCP 80/VCP 130 Vacuum/Pressure Pumps
Pumps Vacuum Diaphragm Pumps
VCP 80 and VCP 130 can be used as vacuum and pressure pumps, and set a standard for efficient vacuum use. Use these pumps as an affordable vacuuming application where corrosion resistance is not required. These pumps can be used for vacuum filtration of aqueous solutions, or aqueous fumes including buffers, but are not recommended for pumping acidic, basic or organic fumes. These VWR® Collection vacuum/pressure pumps are designed for continuous unattended operation, and have two dial gauges with manual regulators that enable you to adjust and easily monitor the required vacuum or pressureMore Product Information
Order Now

SPECIFICATIONS

Model VCP 130 VCP 80
Environmentally Preferable
Flow rate 18 l/min 38 l/min
Vacuum 133 mbar 80 mbar
Final pressure (abs.) 7900 mbar 5200 mbar
Motor output 0,093 kW 0,19 kW
W×D×H 206×224×254 mm 254×191×229 mm
Weight 5,3 kg 5,3 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR