VWR® Peristaltic Pump, Feeding
Supplier: VWR Collection
VWRI181-0590EA 5570 SGD
VWRI181-0590 VWRI181-0591 VWRI181-0592 VWRI181-0593
VWR® Peristaltic Pump, Feeding
Pumps Peristaltic Pumps
VWR® peristaltic pump variants offer a selection of feeding pumps with different pump heads integrated into the pump housing. They cover a broad spectrum of application areas in the laboratory and in industry.
  • Pump heads are made of durable PPS plastic, single - and multi-channel pump heads, are integrated into the pump housing
  • Precise feeding with right-left rotation due to the precisely adjustable motor
  • Operating with brushless DC motor, 110 - 240 VDC, 50/60 Hz
  • Robustness and stability prevent pulsation oscillations and vibrations
  • Control of the
More Product Information