VWR®, Pumps, Peristaltic, Dosing
Supplier: VWR Collection
VWRI181-0594EA 9200 SGD
VWRI181-0594 VWRI181-0595 VWRI181-0596 VWRI181-0597
VWR®, Pumps, Peristaltic, Dosing
Pumps Peristaltic Pumps
VWR® peristaltic pump variants offer a selection of dosing pumps with different pump heads integrated into the pump housing. They cover a broad spectrum of application areas in the laboratory and in industry.
  • Pump heads are made of durable PPS plastic, single- and multi-channel pump heads, integrated to the pump housing
  • Precise dosing with right-left rotation due to the precisely adjustable motor
  • Calibration function
  • Operating with brushless DC motor, 110 to 240 VDC, 50/60 Hz
  • Robustness and stability prevent pulsation oscillations and vibrations
  • Control of
More Product Information