VWR®, Dimroth Condensers with Standard Ground Joints
Supplier: VWR Collection
VWRI201-1409EA 61.7 SGD
VWRI201-1409 VWRI201-1412 VWRI201-1415
VWR®, Dimroth Condensers with Standard Ground Joints
Condensers
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Standard ground cone and socket
These Dimroth condensers are equipped with a double-spiral for effective condensation of vapours. With GL 14 screw thread and 8 mm external diameter tubing connection.